RIE156328966 Rieke Metals, Inc.

2,2,2-trifluoro-2',4',6'-trimethylacetophenone

  • MDL number MFCD00151856
  • Cas Number : 313-56-4
  • Molecular Weight : 216.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6545

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6545
Availability details for
Quantity
  • Availability
  • Add to Cart