RIE156328971 Rieke Metals, Inc.

4-(1-Naphthyl)-4-oxobutyric acid

  • MDL number MFCD00086599
  • Cas Number : 4653-13-8
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/7061b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7061b
Availability details for
Quantity
  • Availability
  • Add to Cart