RIE156328972 Rieke Metals, Inc.

5-(1-Naphthyl)-5-oxovaleric acid

  • MDL number MFCD01320087
  • Cas Number : 59345-41-4
  • Molecular Weight : 242.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7061C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7061C
Availability details for
Quantity
  • Availability
  • Add to Cart