RIE156328979 Rieke Metals, Inc.

7-(2-Naphthyl)-7-oxoheptanoic acid

  • MDL number MFCD01320092
  • Cas Number : 13672-52-1
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7062E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7062E
Availability details for
Quantity
  • Availability
  • Add to Cart