RIE156328980 Rieke Metals, Inc.

8-(2-naphthyl)-8-oxooctanoic acid

  • MDL number MFCD01311642
  • Cas Number : 362669-52-1
  • Molecular Weight : 284.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7062F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7062F
Availability details for
Quantity
  • Availability
  • Add to Cart