RIE156328986 Rieke Metals, Inc.

3,5-Dimethoxybenzoylacetonitrile

  • MDL number MFCD00158877
  • Cas Number : 70988-04-4
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6266

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6266
Availability details for
Quantity
  • Availability
  • Add to Cart