RIE156328989 Rieke Metals, Inc.

6-chloro-1-(3-methoxyphenyl)-1-oxohexane

  • MDL number MFCD07700092
  • Cas Number : 258882-50-7
  • Molecular Weight : 240.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6143E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6143E
Availability details for
Quantity
  • Availability
  • Add to Cart