RIE156328990 Rieke Metals, Inc.

3,5-Dimethyl-4-methoxybenzoylacetonitrile

  • MDL number MFCD02260791
  • Cas Number : 884504-21-6
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6279

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6279
Availability details for
Quantity
  • Availability
  • Add to Cart