RIE156328992 Rieke Metals, Inc.

2-Trifluoromethylbenzoylacetonitrile

  • MDL number MFCD02260792
  • Cas Number : 40018-10-8
  • Molecular Weight : 213.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6280

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6280
Availability details for
Quantity
  • Availability
  • Add to Cart