RIE156328997 Rieke Metals, Inc.

4-Isopropoxybenzoylacetonitrile

  • MDL number MFCD02260796
  • Cas Number : 884504-22-7
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6285

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6285
Availability details for
Quantity
  • Availability
  • Add to Cart