RIE156328999 Rieke Metals, Inc.

3,5-bis(trifluoromethyl)benzoylacetonitrile

  • MDL number MFCD02260797
  • Cas Number : 267880-81-9
  • Molecular Weight : 281.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6286

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6286
Availability details for
Quantity
  • Availability
  • Add to Cart