RIE156329000 Rieke Metals, Inc.

5-fluoro-2-methylbenzoylacetonitrile

  • MDL number MFCD02260799
  • Cas Number : 884504-23-8
  • Molecular Weight : 177.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6288

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6288
Availability details for
Quantity
  • Availability
  • Add to Cart