RIE156329004 Rieke Metals, Inc.

2,4-difluorobenzoylacetonitrile

  • MDL number MFCD02260804
  • Cas Number : 71682-95-6
  • Molecular Weight : 181.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6296

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6296
Availability details for
Quantity
  • Availability
  • Add to Cart