RIE156329006 Rieke Metals, Inc.

4-(2-Fluorophenyl)-4-oxobutyric acid

  • MDL number MFCD01320115
  • Cas Number : 90429-80-4
  • Molecular Weight : 196.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/7069b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7069b
Availability details for
Quantity
  • Availability
  • Add to Cart