RIE156329012 Rieke Metals, Inc.

5-(4-Biphenyl)-5-oxovaleric acid

  • MDL number MFCD01320127
  • Cas Number : 51994-35-5
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7072C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7072C
Availability details for
Quantity
  • Availability
  • Add to Cart