RIE156329015 Rieke Metals, Inc.

8-(4-Biphenyl)-8-oxooctanoic acid

  • MDL number MFCD01311659
  • Cas Number : 362669-53-2
  • Molecular Weight : 310.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7072F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7072F
Availability details for
Quantity
  • Availability
  • Add to Cart