RIE156329016 Rieke Metals, Inc.

2,3-Dimethoxybenzoylacetonitrile

  • MDL number MFCD02260815
  • Cas Number : 199102-80-2
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6307

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6307
Availability details for
Quantity
  • Availability
  • Add to Cart