RIE156329018 Rieke Metals, Inc.

2,5-Dimethoxybenzoylacetonitrile

  • MDL number MFCD02260817
  • Cas Number : 898787-03-6
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6309

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6309
Availability details for
Quantity
  • Availability
  • Add to Cart