RIE156329020 Rieke Metals, Inc.

3,4,5-Triethoxybenzoylacetonitrile

  • MDL number MFCD02260819
  • Cas Number : 267880-91-1
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6314

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6314
Availability details for
Quantity
  • Availability
  • Add to Cart