RIE156329021 Rieke Metals, Inc.

2-Acetylbiphenyl

  • MDL number MFCD00053707
  • Cas Number : 2142-66-7
  • Molecular Weight : 196.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6148

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6148
Availability details for
Quantity
  • Availability
  • Add to Cart