RIE156329026 Rieke Metals, Inc.

5-Cyclohexyl-5-oxovaleric acid

  • MDL number MFCD01320140
  • Cas Number : 15971-96-7
  • Molecular Weight : 198.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7082C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7082C
Availability details for
Quantity
  • Availability
  • Add to Cart