RIE156329028 Rieke Metals, Inc.

2,5-Dimethylbenzoylacetonitrile

  • MDL number MFCD02260824
  • Cas Number : 53882-93-2
  • Molecular Weight : 173.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6323

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6323
Availability details for
Quantity
  • Availability
  • Add to Cart