RIE156329034 Rieke Metals, Inc.

4-Chloro-1-(2-methoxyphenyl)-1-oxobutane

  • MDL number MFCD07700085
  • Cas Number : 40877-17-6
  • Molecular Weight : 212.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6142C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6142C
Availability details for
Quantity
  • Availability
  • Add to Cart