RIE156329038 Rieke Metals, Inc.

4-(3-Bromophenyl)-4-oxobutyric acid

  • MDL number MFCD00672056
  • Cas Number : 62903-13-3
  • Molecular Weight : 257.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7002B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7002B
Availability details for
Quantity
  • Availability
  • Add to Cart