RIE156329856 Rieke Metals, Inc.

8-cyclopropyl-8-oxooctanoic acid

  • MDL number MFCD01311666
  • Cas Number : 898766-91-1
  • Molecular Weight : 198.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7085F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7085F
Availability details for
Quantity
  • Availability
  • Add to Cart