RIE156329859 Rieke Metals, Inc.

9,9-Dimethyl-8-oxodecanoic acid

  • MDL number MFCD01311670
  • Cas Number : 898766-93-3
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7086F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7086F
Availability details for
Quantity
  • Availability
  • Add to Cart