RIE156329863 Rieke Metals, Inc.

8-oxoundecanoic acid

  • MDL number MFCD01320171
  • Cas Number : 91214-06-1
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7090F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7090F
Availability details for
Quantity
  • Availability
  • Add to Cart