RIE156329864 Rieke Metals, Inc.

3-(1,3-Dioxan-2-yl)propionic acid

  • MDL number MFCD07700144
  • Cas Number : 774605-67-3
  • Molecular Weight : 160.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7093

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7093
Availability details for
Quantity
  • Availability
  • Add to Cart