RIE156329871 Rieke Metals, Inc.

3-(2-Oxo-2-phenylethyl)benzoic acid

  • MDL number MFCD07700148
  • Cas Number : 898766-97-7
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7103

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7103
Availability details for
Quantity
  • Availability
  • Add to Cart