RIE156329873 Rieke Metals, Inc.

4-(3,5-dimethylphenyl)-2-methyl-4-oxobutyric acid

  • MDL number MFCD02260864
  • Cas Number : 898767-01-6
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7115

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7115
Availability details for
Quantity
  • Availability
  • Add to Cart