RIE156329874 Rieke Metals, Inc.

4-(1,3-Dioxolan-2-ylmethyl)benzoic acid

  • MDL number MFCD06200651
  • Cas Number : 898767-03-8
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7117A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7117A
Availability details for
Quantity
  • Availability
  • Add to Cart