RIE156329877 Rieke Metals, Inc.

6-(3,4-Dichlorophenyl)-6-oxohexanoic acid

  • MDL number MFCD02260869
  • Cas Number : 898767-10-7
  • Molecular Weight : 275.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7120D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7120D
Availability details for
Quantity
  • Availability
  • Add to Cart