RIE156329879 Rieke Metals, Inc.

8-(3,4-dichlorophenyl)-8-oxooctanoic acid

  • MDL number MFCD02260871
  • Cas Number : 898767-13-0
  • Molecular Weight : 303.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7120F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7120F
Availability details for
Quantity
  • Availability
  • Add to Cart