RIE156329880 Rieke Metals, Inc.

5-(3-bromo-4-methylphenyl)-5-oxovaleric acid

  • MDL number MFCD02260873
  • Cas Number : 898767-16-3
  • Molecular Weight : 285.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7121C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7121C
Availability details for
Quantity
  • Availability
  • Add to Cart