RIE156329882 Rieke Metals, Inc.

7-(3-bromo-4-methylphenyl)-7-oxoheptanoic acid

  • MDL number MFCD03844200
  • Cas Number : 898767-22-1
  • Molecular Weight : 313.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7121E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7121E
Availability details for
Quantity
  • Availability
  • Add to Cart