RIE156329884 Rieke Metals, Inc.

4-(4-Bromo-2-methylphenyl)-4-oxobutyric acid

  • MDL number MFCD02260874
  • Cas Number : 898767-28-7
  • Molecular Weight : 271.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7122B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7122B
Availability details for
Quantity
  • Availability
  • Add to Cart