RIE156329885 Rieke Metals, Inc.

5-(4-bromo-2-methylphenyl)-5-oxovaleric acid

  • MDL number MFCD02260875
  • Cas Number : 898767-31-2
  • Molecular Weight : 285.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7122C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7122C
Availability details for
Quantity
  • Availability
  • Add to Cart