RIE156329886 Rieke Metals, Inc.

6-(4-bromo-2-methylphenyl)-6-oxohexanoic acid

  • MDL number MFCD07700150
  • Cas Number : 898767-34-5
  • Molecular Weight : 299.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7122D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7122D
Availability details for
Quantity
  • Availability
  • Add to Cart