RIE156329887 Rieke Metals, Inc.

7-(4-bromo-2-methylphenyl)-7-oxoheptanoic acid

  • MDL number MFCD07700151
  • Cas Number : 898767-37-8
  • Molecular Weight : 313.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7122E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7122E
Availability details for
Quantity
  • Availability
  • Add to Cart