RIE156329888 Rieke Metals, Inc.

8-(4-bromo-2-methylphenyl)-8-oxooctanoic acid

  • MDL number MFCD07700152
  • Cas Number : 898767-40-3
  • Molecular Weight : 327.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7122F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7122F
Availability details for
Quantity
  • Availability
  • Add to Cart