RIE156329889 Rieke Metals, Inc.

4-(3-bromo-5-methylphenyl)-4-oxobutyric acid

  • MDL number MFCD02260877
  • Cas Number : 898767-43-6
  • Molecular Weight : 271.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7124B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7124B
Availability details for
Quantity
  • Availability
  • Add to Cart