RIE156329896 Rieke Metals, Inc.

6-(2-iodophenyl)-6-oxohexanoic acid

  • MDL number MFCD02260886
  • Cas Number : 898790-71-1
  • Molecular Weight : 332.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7133D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7133D
Availability details for
Quantity
  • Availability
  • Add to Cart