RIE156329897 Rieke Metals, Inc.

7-(2-iodophenyl)-7-oxoheptanoic acid

  • MDL number MFCD02260887
  • Cas Number : 898790-74-4
  • Molecular Weight : 346.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7133E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7133E
Availability details for
Quantity
  • Availability
  • Add to Cart