RIE156329899 Rieke Metals, Inc.

4-(3-Iodophenyl)-4-oxobutyric acid

  • MDL number MFCD02260889
  • Cas Number : 898790-80-2
  • Molecular Weight : 304.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7134B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7134B
Availability details for
Quantity
  • Availability
  • Add to Cart