RIE156329902 Rieke Metals, Inc.

7-(3-iodophenyl)-7-oxoheptanoic acid

  • MDL number MFCD02260892
  • Cas Number : 898790-89-1
  • Molecular Weight : 346.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7134E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7134E
Availability details for
Quantity
  • Availability
  • Add to Cart