RIE156329903 Rieke Metals, Inc.

8-(3-iodophenyl)-8-octanoic acid

  • MDL number MFCD02260893
  • Cas Number : 898790-92-6
  • Molecular Weight : 360.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7134F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7134F
Availability details for
Quantity
  • Availability
  • Add to Cart