RIE156329906 Rieke Metals, Inc.

7-(4-iodophenyl)-7-oxoheptanoic acid

  • MDL number MFCD02260897
  • Cas Number : 49618-09-9
  • Molecular Weight : 346.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7135E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7135E
Availability details for
Quantity
  • Availability
  • Add to Cart