RIE156329907 Rieke Metals, Inc.

8-(4-iodophenyl)-8-octanoic acid

  • MDL number MFCD02260898
  • Cas Number : 898790-98-2
  • Molecular Weight : 360.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7135F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7135F
Availability details for
Quantity
  • Availability
  • Add to Cart