RIE156329908 Rieke Metals, Inc.

4-Oxo-4-(2-trifluoromethylphenyl)butyric acid

  • MDL number MFCD00033285
  • Cas Number : 200725-09-3
  • Molecular Weight : 246.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7139B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7139B
Availability details for
Quantity
  • Availability
  • Add to Cart