RIE156329909 Rieke Metals, Inc.

5-oxo-5-(2-trifluoromethylphenyl)valeric acid

  • MDL number MFCD02260912
  • Cas Number : 502651-48-1
  • Molecular Weight : 260.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7139C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7139C
Availability details for
Quantity
  • Availability
  • Add to Cart